Learn More
Superior Glove KeepKleen™ 5 mil Powder Free Latex Disposable Gloves
Catalog No. p-4939975
Change view
Click to view available options
Size:
Small
Medium
Catalog No. Size
296353523 Small
296353524 Medium
2 options
Catalog No. 296353523
Supplier: Superior Glove Works Ltd RDLPFS
Only null left
Medical exam grade gloves provides better resistance to tearing
  • Provides better touch-sensitivity, dexterity and comfort
  • Beaded cuff-edge for extra strength prevents ripping during donning
  • Textured grip
  • CFIA-approved for food handling
  • White
  • Available in small, medium and large sizes
Dental and Medical Exam, Food Processing, Laboratories, Pharmaceutical, Janitorial.
Specifications
Color White
Disposable Single-use
Latex Free No
Material Latex
Powdered Powder Free
Size Small
Thickness (English) 5 mil
Thickness (Metric) 0.12 mm