Learn More
Superior Glove KeepKleen™ 5 mil Powder Free Latex Disposable Gloves
Description
- Provides better touch-sensitivity, dexterity and comfort
- Beaded cuff-edge for extra strength prevents ripping during donning
- Textured grip
- CFIA-approved for food handling
- White
- Available in small, medium and large sizes
Dental and Medical Exam, Food Processing, Laboratories, Pharmaceutical, Janitorial.
Specifications
Specifications
| Color | White |
| Disposable | Single-use |
| Latex Free | No |
| Material | Latex |
| Powdered | Powder Free |
| Size | Small |
| Thickness (English) | 5 mil |
| Thickness (Metric) | 0.12 mm |