Learn More
Superior Glove™ Endura™ Gauntlet Cuff Cowgrain Leather Gloves
Smooth, top-grain leather gloves provide durability and dexterity along with some resistance to oil and water.
Supplier: Superior Glove™ 375CSILL
Description
- Heavyweight, top-grain cowhide leather
- Grain leather palms provide durability and water-repellency
- Split-leather backs
- Gauntlet cuff
Specifications
| Gray, Natural | |
| Top Grain Cowhide Leather | |
| Large | |
| Gauntlet |
| Specific | |
| General Purpose Gloves | |
| White |