missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ Dexterity™ 13-Gauge Nylon Gloves with Foam PVC Palms

Catalog No. 296356930
Change view
Click to view available options
Size:
10
11
6
7
8
9
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296356930 9
296356932 11
296356931 10
296356929 8
296356927 6
296356928 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 296356930 Supplier Superior Glove™ Supplier No. S13BPVC9
Only null left

Fine-gauge nylon construction is durable and abrasion resistant, while also increasing touch sensitivity. Foam PVC coating on the palm enhances grip capabilities in wet or oily conditions.

  • PVC coating provides wear and abrasion resistance similar to that of nitrile, without the risk of glue adhering to its surface
  • Fine-gauge nylon is durable, while also increasing touch sensitivity
  • Enhanced grip in wet or oily conditions
  • Silicone-free for use in the automotive industry

Specifications

ANSI Abrasion Rating ANSI 3
Construction Seamless Knit
Coating Material PVC
Color Blue
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 9
For Use With (Application) Small Parts Handling, Maintenance, Construction, and Automotive Assembly
Puncture Rating ANSI/ISEA 105-16 Level 2
Series S13BPVC
Yarn Gauge 13
Show More Show Less