Learn More
Epredia™ Superfrost™ Plus Adhesion Control Microscope Slide
Histology Superfrost Slide with designated area for known control sample. Available in Standard, or Extended Tab for automated and manual staining.
$175.93 - $175.93
| Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
|
22-042-941
|
Epredia™
I8432PLUS001 |
Standard Superfrost Tab |
Gross of 144 Each for $175.93
Case of 10 Gross for $1,759.30
|
|
|||||
Description
- Superfrost Plus adhesion coating
- 75 x 25 x 1 mm
- Frosted control box outline for placing control samples
- Available in Standard, or Extended Tab
Specifications
| 144 | |
| Superfrost Plus Adhesion |