Learn More
NOTRAX™ Superfoam™ Revive RS Mat
Superfoam™ Revive RS Mat is a durable rubberized sponge mat designed for extreme ergonomic comfort
$74.12 - $333.61
| Catalog Number | Mfr. No. | Length (English) | Width (English) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Length (English) | Width (English) | Price | Quantity & Availability | ||||
19086046
![]() |
NOTRAX™
425S2024BL |
20 in. | 24 in. |
Each for $74.12
|
|
||||
19086047
![]() |
NOTRAX™
425S2440BL |
24 in. | 40 in. |
Each for $148.34
|
|
||||
19086048
![]() |
NOTRAX™
425S2472BL |
24 in. | 72 in. |
Each for $266.88
|
|
||||
19086049
![]() |
NOTRAX™
425S3640BL |
36 in. | 40 in. |
Each for $222.41
|
|
||||
19086050
![]() |
NOTRAX™
425S3660BL |
36 in. | 60 in. |
Each for $333.61
|
|
||||
Description
- Made from a completely closed cell foam blend
- Does not absorb liquids
- Highly resistant to grease, oils and animal and vegetable fats
- Durable, light weight rubber is easy to pick up and move
- Ergonomically designed to provide high level of rebound without bottoming out
- Non-allergenic, silicone-free compound
- Beveled edges on all 4 sided
Specifications
| Black | |
| 1 Ea. |