missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ Super Heavy Duty 3417 CPE Shoe Covers
Description
CPE, disposable white shoe covers help protect shoes and surrounding environments from dust, dirt, grime, and liquids. They're more durable than polypropylene, so you're protected in tough environments. Whether you're in a cleanroom, a hospital, or working at a commercial site, these durable shoe covers help protect against chemicals, dirt, dust, liquid splash, non-hazardous particulates, and even shallow immersion.
- Water-resistant and come with a strong elastic closure
- One size fits most; fits up to a men's size 12 1/2 shoe
- Can be worn in indoor and outdoor environments
- Possible applications include healthcare, controlled environments, food processing, pharmaceuticals, construction, realtors, movers, contract work, and more
Specifications
Specifications
| Size | X-Large |
| Material | CPE |
| Latex Free | Yes |
| Color | Blue |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Elastic Ankle |
| Sole Type | Standard |
| Packaging Quantity | 300/Cs. |
| Resistance | Dry Particulates, Splash |
| Show More |