missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Super Clean™ (Base-Plate Design) Kits

A unique, point-of-use diffusion proof purification system to purify carrier, fuel, and other gases for GC or GC-MS systems

$1,419.23 - $1,426.10

Specifications

For Use With (Application) Purification of Carrier, Fuel, and Other Gases
For Use With (Equipment) GC or GC-MS Systems
View More Specs

Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
111008374
View Documents Promotion Details
MilliporeSigma™ Supelco™
28878U
Carrier Gas Kit
Each for $1,426.10
Only null left
 
111008040
View Documents Promotion Details
MilliporeSigma™ Supelco™
SU861040
Helium Carrier Gas Kit
Each for $1,419.23
Only null left
 
Description

Description

  • Purifiers remove hydrocarbons, oxygen (color indicated), and moisture (color indicated) to better than 6.0 gas (99.9999%) quality at 2 L/min., independent of the original gas quality
Specifications

Specifications

Purification of Carrier, Fuel, and Other Gases
GC or GC-MS Systems
Videos
SDS
Documents

Documents

Super Clean is a trademark of Scientific Glass Technology B.V.