Learn More
MilliporeSigma™ Supelco™ Super Clean™ (Base-Plate Design) Gas Purifiers
For the Purification and Management of GC Gas Streams
$524.31 - $679.81
Specifications
For Use With (Application) Enhancing The Performance of GC Detectors
For Use With (Equipment) GC Detectors
Quantity 1 Ea.
View More Specs
Products 3
Catalog Number Mfr. No. Type Description Price Quantity & Availability  
Catalog Number Mfr. No. Type Description Price Quantity & Availability  
111009588
View Documents Promotion Details
MilliporeSigma™ Supelco™
SU861025
Dual Trap (Hydrocarbon, Moisture) For Fuel Gas
Each for $524.31
Only null left
111009539
View Documents Promotion Details
MilliporeSigma™ Supelco™
SU861026
Triple Trap (Hydrocarbon, Moisture, Oxygen) For Carrier Gas
Each for $573.58
Only null left
111009287
View Documents Promotion Details
MilliporeSigma™ Supelco™
SU861027
Triple Trap (Hydrocarbon, Moisture, Oxygen) For Helium Carrier Gas
Each for $679.81
Only null left
Description
Description
  • Provides gas at the proper purity level and pressure, based on its intended use
  • The purifiers connect into a base plate in such a manner that gas supply is not interrupted during purifier change-out
  • Purifier can be replaced without shutting down the entire system
  • Eliminates the risk of forcing a slug of air into the instrument after purifier change-out
Specifications
Specifications
Enhancing The Performance of GC Detectors
GC Detectors
1 Ea.