missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Super Bright Complete Staining Buffer, eBioscience

Catalog No. SB440142
Change view
Click to view available options
Quantity:
100 tests
1000 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB440142 100 tests
SB440175 1000 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB440142 Supplier Invitrogen™ Supplier No. LSSB440142

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Super Bright Complete Staining Buffer, eBioscience, is designed for use as a supplement to Flow Cytometry Staining Buffer in immunofluorescent staining protocols of cells in suspension.

Super Bright Complete Staining Buffer is only necessary when using more than one polymer dye-conjugated antibody in the same sample to prevent non-specific polymer interactions, which can result in data appearing under-compensated. Super Bright Complete Staining Buffer is provided in a convenient 5 μL/test format and is compatible with traditional fluorochromes, Brilliant Violet dyes, and standard flow cytometry protocols.

Super Bright Complete Staining Buffer is not compatible with UltraComp eBeads microspheres. If using UltraComp eBeads microspheres as a compensation tool, Flow Cytometry Staining Buffer should be used exclusively for all dilutions and washes during staining of the microspheres.

Super Bright Complete Staining Buffer is a fully compatible replacement for Super Bright Staining Buffer (Cat. Nos. SB-4400-42, SB-4400-75), and it shows decreased background in the 450/40 channel on the violet laser.

Super Bright polymer dyes are sold under license from Becton, Dickinson and Company.

Specifications

Content And Storage 0.5 mL, store at 2°C to 8°C
Quantity 100 tests
Volume (Metric) 0.5 mL
Color Black
Shipping Condition Wet Ice
For Use With (Application) Flow Cytometry
Product Line eBioscience
Product Type Staining Buffer

For Research Use Only. Not for use in diagnostic procedures.