Learn More
3M™ Super Bond Film Tape, 396
Description
- Works well on a wide variety of applications, including repairing and reinforcing high-wear areas of rip-stop nylon, holding polyethylene barrel liners in place, splicing large photographic acetate transparencies, and splicing polyester conveyor belts
- Dispensable and appropriate for outdoor use
Specifications
Specifications
| Color | Transparent |
| Dimensions (L x W x H) | 5.1 cm x 32.9 m x 0.1 mm |
| Width (English) | 2 in. |
| Width (Metric) | 51 mm |