missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-4 Capillary Ferrule, Short Design, 0.53 mm Column I.D.
Description
Characteristics: Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation .Material Composition: flexible graphite. Maximum Temperature: 450°C
Specifications
Specifications
| Type | GC Accessories and Adsorbents |
| Diameter (Metric) Inner | 0.8 mm |
| Packaging Quantity | 10/Pk. |
| Description | Ferrule Type: Short |
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC