Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-2A Packed Column Ferrule, 1/8 in.
Characteristics: Seals at 1/4 turn past fingertight. High reusability. Won't stick to metal or glass. Material Composition: DuPont VESPEL SP-21 (85% polyimide, 15% graphite). Maximum Temperature: 400°C
$518.97
Specifications
Diameter (English) Outer 1/8 in.
Diameter (English) Inner 1/8 in.
View More Specs
Products 1
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
111009297
View Documents Promotion Details
MilliporeSigma™ Supelco™
22472
50/Pk.
Each for $518.97
Only null left
Description
Description
Specifications
Specifications
1/8 in.
1/8 in.
Videos
SDS
Documents
Documents
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC