Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-2A Packed Column Ferrule, 1/4 in.
Characteristics: Seals at 1/4 turn past fingertight. High reusability. Won't stick to metal or glass. Material Composition: DuPont VESPEL SP-21 (85% polyimide, 15% graphite). Maximum Temperature: 400°C
$145.44 - $513.79
Specifications
Diameter (English) Outer 1/4 in.
Diameter (English) Inner 1/4 in.
View More Specs
Products 2
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
111010963
View Documents Promotion Details
MilliporeSigma™ Supelco™
SGMA22481
10/Pk.
Each for $145.44
Only null left
111009296
View Documents Promotion Details
MilliporeSigma™ Supelco™
22471
50/Pk.
Each for $513.79
Only null left
Description
Description
Specifications
Specifications
1/4 in.
1/4 in.
Videos
SDS
Documents
Documents
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC