missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-2 Packed Column Ferrule, 1/4 in.
Characteristics: High reusability.Material Composition: DuPont VESPEL SP-1 (100% polyimide)Maximum Temperature: 350°C
Specifications
| Diameter (English) Outer | 1/4 in. |
|---|---|
| Diameter (English) Inner | 1/4 in. |
| Description | Ferrule Type: Packed Column |
Specifications
| 1/4 in. | |
| 1/4 in. | |
| Ferrule Type: Packed Column |
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC