missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Supeltex™ M-2 Double-Tapered Ferrule, 0.10-0.25 mm to like I.D. Column

Max. Temp.: 350°C. Composition: DuPont VESPEL SP-1 (100% polyimide). Characteristics: High reusability.

Supplier:  MilliporeSigma™ Supelco™ 23797

Catalog No. 111011543


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

0.4 mm
Gas Chromatography (GC)
2/Pk.
Ferrule Type: Double Tapered
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Supeltex is a registered trademark of Sigma-Aldrich Co. LLC