missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Supelpak™ -2 matrix styrene-divinylbenzene, 20-60 mesh

Catalog No. 111001831
Change view
Click to view available options
Quantity:
1000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001831 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001831 Supplier MilliporeSigma™ Supelco™ Supplier No. 21130U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Purified XAD-2

Supelpak™-2 is purified Amberlite™ XAD-2™ that has been cleaned to meet and exceed US EPA-recommended criteria for purity, as outlined in Level I Environmental Assessment Procedures Manual. It is the best resin to use for standard air sampling methods requiring resin tested for background TCO (total chromatographic organics) level. Packaged in glass containers.

Specifications

Particle Size 20/60 Mesh
Packaging Jar
Quantity 1000 g
Surface Area (Metric) ˜300 m2/g
Product Line Supelpak
Density 1.07 g/mL at 25°C (skeletal)(lit.)
CAS 9003-70-7