Learn More
MilliporeSigma™ Supelco™ Supelclean™ LC-8 SPE Tube, Bed Wt. 500 mg
Catalog No. 111010360
Change view
Click to view available options
Volume (Metric):
6 mL
Quantity:
30/Pk.
Catalog No. Volume (Metric) Quantity
111010360 6 mL 30/Pk.
1 options
Catalog No. 111010360 Supplier MilliporeSigma™ Supelco™ Supplier No. 57052
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Specifications
Column Phase LC-8
Column Type SPE
Bed Weight 500 mg
For Use With (Application) Food and Beverages
Product Line Supelclean
Type Solid Phase Extraction Column
Volume (Metric) 6 mL
Endcapped Yes
Material PE Frit
Pore Size 60 Å
Quantity 30/Pk.
Surface Area (Metric) 475 m2/g
Show More Show Less
Supelclean is a trademark of Sigma-Aldrich Co. LLC