missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Supelclean™ ENVI™-Chrom P SPE Tube, Bed Wt. 250 mg

Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.

$420.74 - $593.91

Specifications

Column Phase ENVI-Chrom P
Column Type SPE
Bed Weight 250 mg
For Use With (Application) Food and Beverages
Product Line Supelclean ENVI
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Quantity Price Quantity  
111008957
View Documents
MilliporeSigma™ Supelco™
57224
3 mL 54/Pk. N/A
Only null left
 
111016151
View Documents
MilliporeSigma™ Supelco™
57225U
6 mL 30/Pk. N/A
Only null left
 
Description

Description

Styrene/divinyl benzene co-polymer. Retention Mechanism: Reversed-phase or Adsorption. Sample Matrix Compatibility: Aqueous solutions. Particle Size: 80-160 μm; Spherical Shape; Pore Size: 110- 175 Å; Surface Area: 900 m2/g. Highly crosslinked, neutral, specially cleaned styrenedivinylbenzene resin used to retain hydrophobic compounds with some hydrophilic functionality under reversed phase conditions. Highly resistant to extreme pH conditions. Typical applications include aromatics and phenolic compounds from aqueous sample matrices. Used for priority pollutant phenols from aqueous samples.
Specifications

Specifications

ENVI-Chrom P
250 mg
Supelclean ENVI
Polypropylene
900 m2/g
SPE
Food and Beverages
Solid Phase Extraction Column
20-300 Å
Videos
SDS
Documents

Documents

Supelclean and ENVI are trademarks of Sigma-Aldrich Co. LLC