missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Supelclean™ ENVI-Carb™/NH2 SPE Tube, Bed Wt. 500 mg ENVI-Carb/500 mg LC NH2

Catalog No. 111009761
Change view
Click to view available options
Volume (Metric):
3 mL
6 mL
20 mL
Quantity:
20/Pk.
200/Pk.
30/Pk.
300/Pk.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Quantity
111009761 3 mL 20/Pk.
111005399 6 mL 30/Pk.
111005340 6 mL 300/Pk.
111005407 20 mL 20/Pk.
111005336 20 mL 200/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 111009761 Supplier MilliporeSigma™ Supelco™ Supplier No. 54332U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.

Retention Mechanism: Reversed-phase and anion-exchange. Sample Matrix Compatibility: Organic or aqueous solutions. Dual layer SPE tube that contains both Supelclean ENVI-Carb (upper layer) and LC-NH2 (lower layer) SPE sorbents (separated by PE frit). Developed to offer superior clean up when conducting multi-residue pesticide analysis from food (e.g. agricultural products, meats, etc.). . ENVI-carb has a strong affinity towards planar molecules, and can isolate/remove pigments (e.g., chlorophyll and carotinoids) and sterols commonly present in foods and natural products. Supelclean LC-NH2 is an aminopropyl phase that retains fatty acids, organic acids, and some polar pigments and sugars common in food matrices.

Specifications

Column Phase ENVI-Carb/LC-NH2
Column Type SPE
Bed Weight 500 mg ENVI-Carb/500 mg LC NH2
For Use With (Application) Food and Beverages
Product Line Supelclean ENVI
Type Solid Phase Extraction Column
Volume (Metric) 3 mL
Material Polypropylene
Quantity 20/Pk.

Supelclean and ENVI are trademarks of Sigma-Aldrich Co. LLC