Learn More
MilliporeSigma™ Supelco™ Supelclean™ ENVI-Carb™ II/PSA SPE Tube, Bed Wt. 600 mg PSA/300 mg ENVI-Carb™
Catalog No. 111005412
Change view
Click to view available options
Bed Weight:
600 mg PSA/300 mg ENVI-Carb™-II
Catalog No. Bed Weight
111005412 600 mg PSA/300 mg ENVI-Carb™-II
1 options
Catalog No. 111005412 Supplier MilliporeSigma™ Supelco™ Supplier No. 54058U
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Retention Mechanism: Reversed-phase and anion-exchange. Sample Matrix Compatibility: Organic or aqueous solutions. Dual layer SPE tube that contains both Supelclean ENVI-Carb (upper layer) & PSA (lower layer) SPE sorbents (separated by PE frit). Developed to offer superior clean up when conducting multi-residue pesticide analysis from food (e.g. agricultural products, meats, etc.). ENVI-carb has a strong affinity towards planar molecules, and can isolate/remove pigments (e.g., chlorophyll and carotinoids) and sterols commonly present in foods and natural products. Supelclean PSA is a polymerically bonded, ethylenediamine-N-propyl phase that contains both primary and secondary amines. Supelclean PSA has a strong affinity and high capacity for fatty acids, organic acids, and some polar pigments and sugars. Tested for superior cleanliness using GC-FID and GC-MS.
Specifications
Column Phase PSA/ENVI-Carb-II
Column Type SPE
Bed Weight 600 mg PSA/300 mg ENVI-Carb™-II
For Use With (Application) Food and Beverages
Product Line Supelclean ENVI
Type Solid Phase Extraction Column
Volume (Metric) 6 mL
Material PE Frit
Quantity 30/Pk.
Supelclean and ENVI are trademarks of Sigma-Aldrich Co. LLC