Learn More
MilliporeSigma™ Supelco™ Supelclean™ ENVI™-18 SPE Bulk Packing
Description
Retention Mechanism: Reversed-phase. Sample Matrix Compatibility: Aqueous solutions (drinking, ground, waste water). Polymerically bonded, octadecyl (17% C), endcapped. Excellent for cleaning, extracting and concentrating pollutants from aqueous environmental samples. Higher 17% C loading for increased binding capacities and higher recoveries. Higher carbon loading also offers greater resistance to extreme pH conditions. Used for extracting herbicides, fungicides, and pesticides from waste material.
Specifications
Specifications
| Pore Size | 60 Å |
| Quantity | 100 g |
| Surface Area (Metric) | 475 m2/g |
| Product Line | Supelclean ENVI |
Supelclean and ENVI are trademarks of Sigma-Aldrich Co. LLC