Learn More
MilliporeSigma™ Supelco™ Supel™ Swift HLB SPE 96-Well Plate
Description
- Supel™ Swift HLB is a co-polymer solid phase extraction (SPE) phase composed of both hydrophilic and lipophilic functional groups.
- This phase is ideal for extracting a broad range of compounds from aqueous samples, including food and environmental samples, as well as biological samples (eg. serum and plasma).
- Because of the hydrophilic and lipophilic balanced (HLB) property, the polymer material can selectively retain a wide range of compounds with different polarities and LogP values.
- Traditional HLB SPE uses a five-step process for sample preparation that normally includes the conditioning of sorbents with an organic solvent followed by an equilibration step to return sorbents to sample loading conditions.
- The product design allows for direct sample loading onto the sorbent bed, thereby eliminating the conditioning step altogether.
- This product feature can both save time and simplify sample preparation, which may also serve to reduce sample processing errors.
- The 96-well format is perfect for medium to high-throughput laboratories performing large numbers of samples at a time and is also amenable to automation.
Specifications
Specifications
| Bed Weight | 30 mg |
| Particle Size | 40 to 75 μm |
| Matrix | Polymeric HLB Base Material |
| Pore Size | 80 to 210 Å |
| Quantity | 1 Ea. |
| Surface Area (Metric) | 425 to 500 m2/g |