missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Supel™ QuE Z-Sep/C18 Centrifuge Tube, Shaker Compatible

Catalog No. 111009890
Change view
Click to view available options
Quantity:
50/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111009890 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111009890 Supplier MilliporeSigma™ Supelco™ Supplier No. 55506U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.

Dispersive SPE (dSPE), often referred to as the 'QuEChERS' method (Quick, Easy, Cheap, Effective, Rugged, and Safe), is modern sample prep technique that is becoming increasingly popular in the area of multi-residue pesticide analysis in food and agricultural products. Using the QuEChERS method, food/agricultural samples are first extracted with an aqueous miscible solvent (e.g., acetonitrile) in the presence of high amounts of salts (e.g., sodium chloride and magnesium sulfate) and/or buffering agents (e.g. citrate) to induce liquid phase separation and stabilize acid and base labile pesticides, respectively. Upon shaking and centrifugation, an aliquot of the organic phase is subjected to further cleanup using SPE. Unlike traditional methods using SPE tubes, in dispersive SPE, cleanup is facilitated by mixing bulk amounts of SPE (e.g., Supelclean PSA, ENVI-Carb, and/or Discovery DSC-18) with the extract. After sample cleanup, the mixture is centrifuged and the resulting supernatant can either be analyzed directly or can be subjected to minor further treatment before analysis. Supelco carries a line of vials and centrifuge tubes containing pre-determined amounts of salts and SPE sorbents to support the most common method configurations used today.

Specifications

Column Phase Z-Sep/C18
Column Type SPE
For Use With (Application) Food and Beverages
For Use With (Equipment) Most QuEChERS Shakers
Product Line Supel
Type Solid Phase Extraction Column
Volume (Metric) 15 mL
Quantity 50/Pk.

Supel is a trademark of Sigma-Aldrich Co. LLC