missing translation for 'onlineSavingsMsg'
Learn More

Supel™ QuE Z-Sep Bulk, Sold by MilliporeSigma™ Supelco™

Enhances sample cleanup for complex matrices. Effectively removes more fat and color from sample extracts than traditional phases for QuEChERS LC-MS methods.

Supplier:  Merck Emd Millipore 55299U

 View more versions of this product

Catalog No. 111003811


Only null left
Explore available promotions

Description

Description

  • By eliminating problematic matrix interferences, Z-Sep products provide more robust LC-MS and GC-MS methods
  • This proprietary technology can replace C18 and PSA ised in current methods without additional method development
  • Dispersive SPE (dSPE), often referred to as the QuEChERS method, is a modern sample prep technique
  • dSPE is becoming increasingly popular in the area of multi-residue pesticide analysis in food and agricultural products
  • In dispersive SPE cleanup is facilitated by mixing bulk amounts of SPE (e.g., Supelclean PSA, ENVI-Carb, and/or Discovery DSC-18) with the extract
  • After sample cleanup, the mixture is centrifuged and the resulting supernatant can either be analyzed directly or can be subjected to minor further treatment before analysis
  • A line of vials and centrifuge tubes containing pre-determined amounts of salts and SPE sorbents are available to support the most common method configurations used today
Specifications

Specifications

50 μm
Silica Gel Base Material
480 m2/g
Z-Sep+ Bulk
70 Å
20 g
Supel
Cleanup of Samples Containing > 15% Fat
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Certain uses of this product may be covered by the claims of one or more pending patent applications. For licensing inquiries please contact techserv@sial.com