Learn More
Sunflower Seed Oil, Spectrum™ Chemical
S1929, 8001-21-6
$3320.91 - $3320.91
Chemical Identifiers
CAS 8001-21-6
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18607190
View Documents
Spectrum Chemical Mfg Cor
S192920LT
20 L Steel Can
Each for $3,320.91
Only null left
Description
Description
Spectrum™ Chemical Sunflower Seed Oil is, as its name suggests, the oil of sunflower seeds that is extracted by the compression of the seeds. This non-volatile oil is most commonly used in food and as a frying oil. It is also used in cosmetics as an emollient. It is also mixed with diesel to run diesel combustion engines.
Specifications
Chemical Identifiers
8001-21-6
Specifications
8001-21-6
Steel Can
188-194
Reagent
1
20 L
0.914-0.924
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'