Learn More
Waters Limited SunFire C8 Sentry Guard Cartridge, 100, 3.5 m, 2.1 mm X 10 mm, 2/pk
Catalog No. 50781299
Encompass
SunFire C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to SunFire C18.
Catalog No. 50781299 Supplier Waters Limited Supplier No. 186002708
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
SunFire C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to SunFire C18.