missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SUN-SRi™ Luer-LOCK Syringes

Catalog No. 22034510
Change view
Click to view available options
Volume (Metric):
3 mL
10 mL
Sterility:
Non-sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility
22034510 10 mL Non-sterile
14823396 3 mL Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22034510 Supplier Thermo Scientific™ Supplier No. CMD400 014
Only null left

Offered in a range of sizes, from 2mL to 20mL, sterile and non-sterile.

  • Chemically resistant, inert polypropylene and polyethylene
  • No rubber plunger seals or silicone lubricants that may cause sample contamination
  • Bulk packaged, non sterile-no paper wrap waste
  • Luer-LOCK™ connection

Specifications

Material Plastic
Product Type Luer-lok Syringe
Disposable Single-use
Graduations 0.5 mL
Volume (Metric) 10 mL
Needle Point Style Centered
Connection Type Luer Lock
Sterility Non-sterile