Learn More
Sun Spotter Telescope DFS Item
View the brilliant light of the sun safely with this wooden, folded-path Keplerian telescope.
Supplier:  Science First LT70
Catalog No. S01384
Only null left
Explore available promotions
Description
Description
Series of mirrors projects 3.25 in. (8.26cm) solar image through a powerful objective lens and onto a 5 in. (12.7cm) white viewing screen.
  • Curved cradle allows alignment with the sun in seconds
  • Convenient, easy to set up, lightweight and kid friendly
  • Use to view the sun, eclipses and transits and for daily record keeping
  • Track sunspots as they appear, move and vanish
  • Useful for group viewing; allows observers to simultaneously see the sun's disk and trace changing sunspot positions.
Specifications
Specifications
Use to view the sun, eclipses and transits and for daily record keeping
Sun Spotter Telescope
Product Suggestions
Product Suggestions