Learn More
Justrite™ Sump-to-Sump™ Drain Kit for EcoPolyBlend™ Accumulation Centers, Stainless Steel Clips, Grommets, Transfer Tube
Catalog No. 296352270
Change view
Click to view available options
For Use With (Equipment):
EcoPolyBlend Accumulation Centers
Catalog No. For Use With (Equipment)
296352270 EcoPolyBlend Accumulation Centers
1 options
Catalog No. 296352270 Supplier Justrite™ Supplier No. 28927
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Sump-to-Sump™ Drain Kit for EcoPolyBlend™ Accumulation Centers, Stainless Steel Clips, Grommets, Transfer Tube is an accessory to help facilitate the use of our accumulation centers.
Use for applications requiring a continuous sump and for full compliance with regulations when sump capacities reach 71 gal. (270 L). Drain kit includes one pair stainless-steel joining clips, one pair rubber grommets and polytetrafluoroethylene transfer tube. It allows you to join centers together, on site, using unites 4-drum and smaller, to reach needed capacity. By joining centers together with the patented Sump-to-Sump Drain™ kit for a total 66-gallon sump capacity, you will meet EPA and UFC requirements.
Specifications
For Use With (Equipment) EcoPolyBlend Accumulation Centers