missing translation for 'onlineSavingsMsg'
Learn More

Niacinamide, USP, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18604795
Change view
Click to view available options
Quantity:
500 g
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604795 25 kg Fiber Drum
18604792 500 g Poly Bottle
18604794 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604795 Supplier Spectrum Chemical Mfg Cor Supplier No. NI10525KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

NI105, 98-92-0, C6H6N2O

Spectrum™ Chemical Niacinamide, USP is a vitamin supplement. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 98-92-0
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
InChI Key DFPAKSUCGFBDDF-UHFFFAOYSA-N
IUPAC Name pyridine-3-carboxamide
SMILES NC(=O)C1=CC=CN=C1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Melting Range 128-131°C
Packaging Fiber Drum
Quantity 25 kg
Percent Purity 98.5 to 101.5%
Grade USP