missing translation for 'onlineSavingsMsg'
Learn More

Sumatriptan Succinate 98.0+%, TCI America™

Catalog No. S0851100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0851100MG 100 mg
S08511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0851100MG Supplier TCI America Supplier No. S0851100MG

Chemical Identifiers

CAS 103628-48-4
Molecular Formula C18H27N3O6S
Molecular Weight (g/mol) 413.489
MDL Number MFCD00902856
InChI Key PORMUFZNYQJOEI-UHFFFAOYSA-N
Synonym 3-[2-(Dimethylamino)ethyl]-1H-indol-5-yl-N-methylmethanesulfonamide Succinate
PubChem CID 59772
ChEBI CHEBI:64359
IUPAC Name butanedioic acid;1-[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]-N-methylmethanesulfonamide
SMILES CNS(=O)(=O)CC1=CC2=C(C=C1)NC=C2CCN(C)C.C(CC(=O)O)C(=O)O

Specifications

Melting Point 167°C
Color White-Yellow
pH 4.8 (1%aq)
Quantity 100 mg
Formula Weight 413.49
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sumatriptan Succinate
RTECSNumber EJ9940000
TSCA No