missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sulindac Sulfide

CAS: 32004-67-4 | C20H17FO2S, C20H17FO2S | 340.41 g/mol

Supplier:  Thermo Scientific Chemicals J62104MC

Catalog No. AAJ62104MC


Only null left

Description

Description

It is also shown to inhibit growth and induce apoptosis in human prostate cancer cells through a COX-1 and COX-2 independent mechanism. Sulindac sulfide is an analgesic that has antiproliferative and apoptotic effects. It inhibits the expression and activity of cyclooxygenase-2 in human colon cancer cells, and reduces tumor burden in adenomatous polyposis patients. Human endothelial cells, HMEC-1 were treated with Sulindac sulfide and the effect on cell survival was studies by MTT assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 250-892-2

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only