missing translation for 'onlineSavingsMsg'
Learn More

Sulfuryl chloride, 1.0M solution in dichloromethane, AcroSeal™

CAS: 7791-25-5 | Cl2O2S | 134.96 g/mol

$118.52 - $118.52

Specifications

Chemical Name or Material Sulfuryl chloride
Name Note 1.0M Solution in Dichloromethane
CAS 7791-25-5,75-09-2
Molecular Formula Cl2O2S
Linear Formula SO2Cl2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC378151000
View Documents
Thermo Scientific Chemicals
378151000
100 mL AcroSeal™ Glass bottle
Each for $118.52
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Specifications

Sulfuryl chloride
7791-25-5,75-09-2
SO2Cl2
15, 9110
YBBRCQOCSYXUOC-UHFFFAOYSA-N
sulfuryl dichloride
24648
134.97
Clear pale yellow solution
1.3520g/mL
1.0M Solution in Dichloromethane
Cl2O2S
MFCD00011451
sulfuryl chloride, sulfonyl chloride, sulphuryl dichloride, sulphuryl chloride, sulfonyl dichloride, sulfuric dichloride, sulfuric oxychloride, sulfurylchlorid, caswell no. 816, sulfurylchloride
ClS(Cl)(=O)=O
134.96
CHEBI:29291
1M, exact strength on the certificate of analysis
1.352
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only