missing translation for 'onlineSavingsMsg'
Learn More

Sulfurous Acid (Certified ACS), Fisher Chemical

Catalog No. a3071
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A3071 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A3071 Supplier Thermo Fisher Scientific Supplier No. FLA3071
Only null left

Chemical Identifiers

CAS 7782-99-2
Molecular Formula H2O3S
Molecular Weight (g/mol) 82.073
MDL Number MFCD00011335
InChI Key LSNNMFCWUKXFEE-UHFFFAOYSA-N
Synonym sulphurous acid, sulfur dioxide solution, schweflige saure german, h2so3, unii-j1p7893f4j, schweflige saure, acide sulfureux, acido sulfuroso, schweflige saeure, trioxosulfuric acid
PubChem CID 1100
ChEBI CHEBI:48854
IUPAC Name sulfurous acid
SMILES OS(=O)O

Specifications

pH 2.0
Density 1.03g/cm³
Heavy Metals (as Pb) 2ppm max.
Identification Pass Test
Packaging Amber Glass Bottle
Quantity 1 L
Physical Form Liquid
UN Number 1833
Percent Purity ≥6.0%
Grade Certified ACS
Chemical Name or Material Sulfurous Acid
Show More Show Less
DOTInformation Class 8: Corrosive