missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfurous Acid, 6% ACS Reagent, Reagents
Sulfurous Acid, 6% ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C111550025Y
Specifications
| <7 | |
| Colorless | |
| 35°C | |
| 1.03 | |
| H2O3S | |
| Sulfur dioxide solution, Sulphurous Acid | |
| OS(O)=O | |
| 82.07 |
| Sulfurous Acid, 6% ACS Reagent | |
| 7782-99-2,7732-18-5 | |
| Glass Bottle | |
| 2.5 L | |
| Liquid | |
| LSNNMFCWUKXFEE-UHFFFAOYSA-N | |
| sulfurous acid | |
| ACS |
Chemical Identifiers
| 7782-99-2 | |
| 82.07 | |
| Sulfur dioxide solution, Sulphurous Acid | |
| OS(O)=O |
| H2O3S | |
| LSNNMFCWUKXFEE-UHFFFAOYSA-N | |
| sulfurous acid |