missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, Ultratrace, 93-98%, Spectrum™ Chemical

Catalog No. p-7166272
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
PTFE Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614299 500 mL PTFE Bottle
18614298 1 L PTFE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614299 Supplier Spectrum Chemical Mfg Cor Supplier No. S1072500MLTF
Only null left

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Max % 1
Packaging PTFE Bottle
Quantity 500 mL
Percent Purity 93 to 98%
Grade High Purity Solvent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.