missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, TraceSELECT™ Ultra, ≥95% (T), Honeywell Fluka™

Catalog No. p-7120856
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly bottle
Catalog No. Quantity Packaging
60047546 500 mL Poly bottle
60047544 1 L Poly bottle
2 options

Catalog No. 60047546

Supplier: Honeywell Inc 77239500ML

Only null left

TraceSELECT™ Ultra, for ultratrace analysis,≥95%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH <1
Melting Point -18°C
Boiling Point 203°C
Density 1.66g/cm3 at 20°C
Odor Odorless
Packaging Poly bottle
Vapor Pressure 1mmHg (146°C)
Quantity 500 mL
Physical Form Liquid
Linear Formula H2SO4
UN Number UN1830
Beilstein 2037554
Formula Weight 98.08g/mol
Percent Purity ≥95% (T)
Grade TraceSELECT™ Ultra
Chemical Name or Material Sulfuric acid
Show More Show Less
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
DOTInformation 8 / PGII
RTECSNumber 231-639-5