missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid (TraceMetal™ Grade), Fisher Chemical™

Catalog No. p-20557
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
HDPE Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A510P500 500 mL HDPE Bottle
A510P212 2.5 L HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A510P500 Supplier Seastar Chemicals Inc Supplier No. S010301TFIQ03
Only null left
  • Certificate of Lot Analysis included with each shipment.
  • Ideal for analysis by IC, GF or Flame AA, ICP-OES and ICP-MS.
  • Evaluated for 65 metals at <1 ppb concentrations. Manufactured, purified, and packaged in a Class1000 clean room.
  • Packaged in non-breakable polypropylene bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture available on COA. 45mm bottle neck size.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0.3
Color Colorless to Brown
Melting Point 10°C
Boiling Point 290°C
Density 1.84g/cm³
Packaging HDPE Bottle
Vapor Pressure <0.001 mmHg at 20°C
Certificate of Analysis Certificate of Lot Analysis included with each shipment.
Quantity 500 mL
Physical Form Liquid
UN Number 1830
Formula Weight 98.08
Grade TraceMetal
Chemical Name or Material Sulfuric Acid
Show More Show Less
DOTInformation DOT Class 8, : Corrosive