missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, TraceGrade, 94-98%, Spectrum™ Chemical

Catalog No. 18614297
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614297 500 mL Poly Bottle
18614296 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614297 Supplier Spectrum Chemical Mfg Cor Supplier No. S1136500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 500 mL
Percent Purity 94 to 98%
Grade High Purity Solvent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.