missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid Solution, 49 to 51%, For HPLC, Honeywell Fluka™
for HPLC, 49-51%
$1175.38 - $1175.38
Chemical Identifiers
| CAS | 7664-93-9 |
|---|---|
| Molecular Formula | H2O4S |
| Molecular Weight (g/mol) | 98.07 |
| MDL Number | MFCD00064589 |
| InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
| Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
| PubChem CID | 1118 |
| ChEBI | CHEBI:26836 |
| IUPAC Name | sulfuric acid |
| SMILES | OS(O)(=O)=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
Specifications
| Acidic | |
| 7664-93-9 | |
| 1.403g/mL at 20°C | |
| Glass bottle | |
| H2O4S | |
| MFCD00064589 | |
| 2037554 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 98.08g/mol | |
| Sulfuric acid solution |
| Colorless | |
| 110°C | |
| Odorless | |
| 500 mL | |
| H2SO4 | |
| UN2796 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| 49 to 51% |
Safety and Handling
P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
DOTInformation : 8 / PGII