missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid solution, 0.25M, Reag. Ph. Eur., Solstice

Catalog No. 60046575
Change view
Click to view available options
Quantity:
5 L
10 L
4 x 5 L
Packaging:
Plastic Bottle
VOLPAC™ CONTAINER
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046575 4 x 5 L Plastic Bottle
6001609 5 L Plastic Bottle
6001607 10 L VOLPAC™ CONTAINER
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60046575 Supplier Honeywell Inc Supplier No. 353554X5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Volumetric, Reag. Ph. Eur., 0.25 M H2SO4 (0.5N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Packaging Plastic Bottle
Linear Formula H2SO4
Quantity 4 x 5 L
UN Number UN3264
Formula Weight 98.08
Grade Reagent Ph. Eur.
Chemical Name or Material Sulfuric acid solution, volumetric, 0.25 M H2SO4 (0.5N)
DOTInformation 8 / PGIII