missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, NF, 95-98%, Spectrum™ Chemical

Catalog No. 18607168
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607168 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 18607168 missing translation for 'mfr' Spectrum Chemical Mfg Cor missing translation for 'supplierNo' S1731500MLGL
Only null left

S1731, 7664-93-9, H2SO4

Spectrum™ Chemical Sulfuric Acid, NF is used as an acidifying agent in pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 95 to 98%
Grade NF