missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, NF, 95-98%, Spectrum™ Chemical

Catalog No. p-7158343
Change view
Click to view available options
Quantity:
2.5 L
20 L
4 x 2.5 L
Packaging:
Glass Bottle
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607171 2.5 L Glass Bottle
18607173 4 x 2.5 L Glass Bottle
18607175 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18607171 Supplier Spectrum Chemical Mfg Cor Supplier No. S17312.5LTGL
Only null left

S1731, 7664-93-9, H2SO4

Spectrum™ Chemical Sulfuric Acid, NF is used as an acidifying agent in pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Max % 1
Packaging Glass Bottle
Quantity 2.5 L
Percent Purity 95 to 98%
Grade NF
Chemical Name or Material Sulfuric Acid