missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, For the determination of nitrogen, 97.5%, MilliporeSigma™ Supelco™

Catalog No. p-7193462
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
111019607 500 mL Poly Bottle
111018399 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019607 Supplier Merck Emd Millipore Supplier No. 84727500ML
Only null left
Sulfuric acid can be produced by dry distillation of iron sulfate heptahydrate, copper sulfate pentahydrate. When these compounds were heated they decompose to form iron and copper oxides eliminating water and sulphur trioxide which combine together to form dilute solution of sulphuric acid. It is suitable as dehydrating agent, catalyst, active reactant in chemical industry. It is also suitable to control pH of saline solutions to strong fuming acids.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Clear
Boiling Point ˜290°C
Density 1.840 g/mL (at 25°C)
Packaging Poly Bottle
Vapor Pressure 1 mmHg (146°C)
Quantity 500 mL
Linear Formula H2SO4
Formula Weight 98.08
Percent Purity ≥97.5%
RTECSNumber WS5600000