missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid (For Babcock Milk Test), Fisher Chemical™

Catalog No. p-20538
Change view
Click to view available options
Quantity:
2.5 L
4 L
Packaging:
Poly Bottle
PPQ Resin Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SA174212 2.5 L PPQ Resin Bottle
SA1744 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SA174212 Supplier Thermo Fisher Scientific Formerly Fairlawn Supplier No. SA174 212
Only null left

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0.3
Color Colorless to Brown
Melting Point 10°C
Boiling Point 290°C
Density 1.84g/cm³
Identification Pass Test
Packaging PPQ Resin Bottle
Specific Gravity 1.82 to 1.83 (at 25°C)
Vapor Pressure <0.001 mmHg at 20°C
Assay Percent Range 90.5 to 93.0 %
Quantity 2.5 L
Physical Form Liquid
UN Number 1830
Formula Weight 98.08
Percent Purity 90.5 to 93.0%
Chemical Name or Material Sulfuric Acid, For Babcock Milk Test
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes severe burns by all exposure routes. Reacts violently with water. Contact with combustible material may cause fire. Exposure to strong inorganic mists containing sulfuric acid may cause cancer by inhalation. Hygroscopic. Use personal protective equipment. Do not allow contact with water. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:3
Flammability:0
Instability:2
DOTInformation DOT Class 8, : Corrosive