missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, extra pure, fuming, 20-30% free SO3

CAS: 8014-95-7 | H2O7S2 | 178.13 g/mol

Supplier:  Thermo Scientific Chemicals 419970025

 View more versions of this product

Catalog No. AC419970025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sulfuric acid
2.0°C
138.0°C
Glass bottle
1.92
H2O7S2
H2SO4·SO3
15, 9104
Solubility in water: miscible. Other solubilities: soluble in ethanol
O=S(=O)=O.OS(O)(=O)=O
178.13
178.14
Extra Pure
Brown to Colorless
8014-95-7,7446-11-9
1.9200g/mL
0.01% max.
2.5 kg
Liquid
MFCD00137792
4-bromo-n-2-methylphenyl benzenesulfonamide, 4-bromo-n-o-tolyl-benzenesulfonamide, 4-bromobenzenesulfon-o-toluide, 4-bromo-2'-methylbenzenesulfonanilide, 4-bromo-n-o-tolyl benzenesulfonamide, 4-bromo-n-2-methylphenyl benzenesulfonamide #
HIFJUMGIHIZEPX-UHFFFAOYSA-N
4-bromo-N-(2-methylphenyl)benzenesulfonamide
571910
20 to 30% (free SO3)

Chemical Identifiers

8014-95-7
178.13
HIFJUMGIHIZEPX-UHFFFAOYSA-N
H2O7S2
MFCD00137792
O=S(=O)=O.OS(O)(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

RTECSNumber : WS5605000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only