missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, Electronic/Cleanroom Grade, 95-97%, Spectrum™ Chemical
S1742, 7664-93-9, H2SO4
$1154.33 - $1154.33
Chemical Identifiers
| CAS | 7664-93-9 |
|---|---|
| Molecular Formula | H2O4S |
| Molecular Weight (g/mol) | 98.07 |
| MDL Number | MFCD00064589 |
| InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
| IUPAC Name | sulfuric acid |
| SMILES | OS(O)(=O)=O |
Description
Spectrum™ Chemical Sulfuric Acid, Electronic/Cleanroom Grade is an extremely corrosive acid that comes as a yellowish slightly viscous liquid. It is soluble in water and is a diprotic acid. It has very strong corrosive, dehydrating and oxidizing properties as well as being hygroscopic. Spectrumâs ELECTRONIC/CLEANROOM GRADE products are suitable for use in electronic component manufacturing. These materials meet or exceed the specifications as listed in Reagent Chemicals published by the American Chemical Society.
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| OS(O)(=O)=O |
| H2O4S | |
| MFCD00064589 | |
| sulfuric acid |
Specifications
| 7664-93-9 | |
| Poly Bottle | |
| H2O4S | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 95 to 97% | |
| Sodium Sulfathiazole, Sesquihydrate |
| 1 | |
| 1 gal. | |
| MFCD00064589 | |
| OS(O)(=O)=O | |
| 98.07 | |
| Electronic/Cleanroom |