missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, Certified, 20.0% (v/v) ±0.5%, LabChem™
Supplier: LabChem LC256074
Specifications
| Sulfuric Acid, (1+4) | |
| 7664-93-9,7732-18-5 | |
| 29.15,70.85 | |
| Poly Bottle | |
| Traceable to NIST | |
| 4 L | |
| Liquid | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 98.08 | |
| Certified |
| Colorless | |
| 1.21g/mL | |
| Passes Test | |
| 1.21g/mL | |
| Sulfur compounds; Thermal decomposition generates: Corrosive vapors | |
| H2O4S | |
| H2SO4 | |
| UN2796 | |
| Soluble in water | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| 20.0% ±0.5% (v/v) |
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse
Danger
Recommended Storage : Room Temperature