missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, Certified, 0.0200N ±0.0002N (0.01M), LabChem™
Supplier: LabChem LC256602
Specifications
| Sulfuric Acid, 0.02N (0.01M) | |
| 7664-93-9,7732-18-5 | |
| 1g/mL | |
| Passes Test | |
| 1g/mL | |
| Sulfur compounds | |
| 1cSt | |
| Liquid | |
| MFCD00064589 | |
| Soluble in water | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| Certified |
| Colorless | |
| 0.0200N ± 0.0002N | |
| 99.9,0.1 | |
| Poly Bottle | |
| Traceable to NIST | |
| 1 L | |
| H2O4S | |
| H2SO4 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 98.08 |
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/at
Recommended Storage : Room Temperature